missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Phenylpiperidine 98.0+%, TCI America™
$182.68 - $676.65
Chemical Identifiers
| CAS | 3973-62-4 |
|---|---|
| Molecular Formula | C11H15N |
| Molecular Weight (g/mol) | 161.25 |
| MDL Number | MFCD00804860 |
| InChI Key | NZYBILDYPCVNMU-UHFFFAOYNA-N |
| PubChem CID | 107207 |
| IUPAC Name | 3-phenylpiperidine |
| SMILES | C1CNCC(C1)C1=CC=CC=C1 |
Chemical Identifiers
| 3973-62-4 | |
| 161.25 | |
| NZYBILDYPCVNMU-UHFFFAOYNA-N | |
| 3-phenylpiperidine |
| C11H15N | |
| MFCD00804860 | |
| 107207 | |
| C1CNCC(C1)C1=CC=CC=C1 |
Specifications
| 3973-62-4 | |
| Yellow | |
| C11H15N | |
| 1 g | |
| C1CNCC(C1)C1=CC=CC=C1 | |
| 161.25 | |
| 161.25 | |
| Liquid |
| 15°C | |
| 126°C | |
| MFCD00804860 | |
| NZYBILDYPCVNMU-UHFFFAOYNA-N | |
| 3-phenylpiperidine | |
| 107207 | |
| ≥98.0% (T) | |
| 3-Phenylpiperidine |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator