accessibility menu, dialog, popup

UserName

HN

CAS RN 3973-62-4

IUPAC Name: 3-phenylpiperidine
Synonyms: 3-phenyl-piperidine
Molecular Weight (g/mol): 161.25
Molecular Formula: C11H15N
InChi Key: NZYBILDYPCVNMU-UHFFFAOYNA-N
SMILES: C1CNCC(C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results