missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Phenylpiperidine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 15°C |
| Color | Yellow |
| Boiling Point | 126°C |
| Quantity | 1 g |
| Formula Weight | 161.25 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Phenylpiperidine |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |