missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Phenylphthalide 98.0+%, TCI America™
Supplier: TCI America P180625G
Specifications
| 3-Phenylphthalide | |
| 118°C | |
| C14H10O2 | |
| 25 g | |
| SQFMIHCARVMICF-UHFFFAOYNA-N | |
| 3-phenyl-3H-2-benzofuran-1-one | |
| 95365 | |
| ≥98.0% (GC) |
| 5398-11-8 | |
| White | |
| MFCD00023099 | |
| 3-phenylphthalide, 3-phenyl phthalide, phthalide, 3-phenyl, 3-phenyl-2-benzofuran-1 3h-one, 3-phenyl-1 3h-isobenzofuranone, 3-phenyl-1,3-dihydro-2-benzofuran-1-one, 1 3h-isobenzofuranone, 3-phenyl, 3-phenyl-3-hydroisobenzofuran-1-one, phenylphthalid, acmc-1ak9n | |
| C1=CC=C(C=C1)C2C3=CC=CC=C3C(=O)O2 | |
| 210.23 | |
| 210.23 | |
| Crystalline Powder |
Chemical Identifiers
| 5398-11-8 | |
| 210.23 | |
| SQFMIHCARVMICF-UHFFFAOYNA-N | |
| 95365 | |
| C1=CC=C(C=C1)C2C3=CC=CC=C3C(=O)O2 |
| C14H10O2 | |
| MFCD00023099 | |
| 3-phenylphthalide, 3-phenyl phthalide, phthalide, 3-phenyl, 3-phenyl-2-benzofuran-1 3h-one, 3-phenyl-1 3h-isobenzofuranone, 3-phenyl-1,3-dihydro-2-benzofuran-1-one, 1 3h-isobenzofuranone, 3-phenyl, 3-phenyl-3-hydroisobenzofuran-1-one, phenylphthalid, acmc-1ak9n | |
| 3-phenyl-3H-2-benzofuran-1-one |
Safety and Handling
RTECSNumber : TI3800000
TSCA : No