missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylphthalide 98.0+%, TCI America™

Catalog No. p-7135263
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18065G 5 g
P180625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18065G Supplier TCI America Supplier No. P18065G
Only null left

Chemical Identifiers

CAS 5398-11-8
Molecular Formula C14H10O2
Molecular Weight (g/mol) 210.23
MDL Number MFCD00023099
InChI Key SQFMIHCARVMICF-UHFFFAOYNA-N
Synonym 3-phenylphthalide, 3-phenyl phthalide, phthalide, 3-phenyl, 3-phenyl-2-benzofuran-1 3h-one, 3-phenyl-1 3h-isobenzofuranone, 3-phenyl-1,3-dihydro-2-benzofuran-1-one, 1 3h-isobenzofuranone, 3-phenyl, 3-phenyl-3-hydroisobenzofuran-1-one, phenylphthalid, acmc-1ak9n
PubChem CID 95365
IUPAC Name 3-phenyl-3H-2-benzofuran-1-one
SMILES C1=CC=C(C=C1)C2C3=CC=CC=C3C(=O)O2

Specifications

Melting Point 118°C
Color White
Quantity 5 g
Formula Weight 210.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Phenylphthalide
RTECSNumber TI3800000
TSCA No