missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyllactic acid, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111001866
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001866 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001866 Supplier Merck Emd Millipore Supplier No. 9226725MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 828-01-3
Linear Formula C6H5 CH2 CH(OH)COOH
MDL Number MFCD00065928
Synonym alpha-Hydroxyhydrocinnamic acid; beta-Phenyllactic acid; 2-Hydroxy-3-phenylpropionic acid
Type Analytical Standard
Quantity 25 mg
Formula Weight 166.17
Percent Purity ≥98% (GC)
Grade Analytical Standard