missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylimidazo[1,5-a]pyridine 98.0+%, TCI America™

Catalog No. P18385G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18385G 5 g
P183825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18385G Supplier TCI America Supplier No. P18385G
Only null left

Chemical Identifiers

CAS 35854-46-7
Molecular Formula C13H10N2
Molecular Weight (g/mol) 194.237
InChI Key BLGGTXOVNVHSSV-UHFFFAOYSA-N
PubChem CID 606670
IUPAC Name 3-phenylimidazo[1,5-a]pyridine
SMILES C1=CC=C(C=C1)C2=NC=C3N2C=CC=C3

Specifications

Melting Point 110°C
Boiling Point 140°C
Quantity 5 g
Formula Weight 194.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Phenylimidazo[1,5-a]pyridine