accessibility menu, dialog, popup

UserName

NN

CAS RN 35854-46-7

IUPAC Name: 3-phenylimidazo[1,5-a]pyridine
Synonyms: 3-phenylimidazo[1,5-a]pyridine
Molecular Weight (g/mol): 194.24
Molecular Formula: C13H10N2
InChi Key: BLGGTXOVNVHSSV-UHFFFAOYSA-N
SMILES: C1=C2C=CC=CN2C(=N1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results