missing translation for 'onlineSavingsMsg'
Learn More

3-(Phenylamino)propyltrimethoxysilane, 96%

Catalog No. p-7046935
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1685014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1685014 Supplier Thermo Scientific Chemicals Supplier No. L1685014

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3068-76-6 | C12H21NO3Si | 255.389 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3068-76-6
Molecular Formula C12H21NO3Si
Molecular Weight (g/mol) 255.389
MDL Number MFCD00053673
InChI Key KBJFYLLAMSZSOG-UHFFFAOYSA-N
Synonym n-3-trimethoxysilyl propyl aniline, trimethoxy 3-phenylamino propyl silane, n-phenylaminopropyltrimethoxysilane, unii-7au1h1fv6a, benzenamine, n-3-trimethoxysilyl propyl, n-phenyl-3-aminopropyltrimethoxysilane, 7au1h1fv6a, n-3-trimethoxysilylpropyl aniline, 3-phenylamino propyltrimethoxysilane
PubChem CID 76476
IUPAC Name N-(3-trimethoxysilylpropyl)aniline
SMILES CO[Si](CCCNC1=CC=CC=C1)(OC)OC

Specifications

Boiling Point 310°C
Quantity 25 g
Flash Point 165°C (329°F)
Odor Weak
Refractive Index 1.5055
Sensitivity Moisture sensitive
Density 1.07
Linear Formula C6H5NH(CH2)3Si(OCH3)3
UN Number UN3267
Beilstein 8620045
Formula Weight 255.39
Percent Purity 96%
Chemical Name or Material 3-(Phenylamino)propyltrimethoxysilane
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H314-H318-H351-H373
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
EINECSNumber 221-328-2
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only