missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Phenylacrylaldehyde oxime, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals BTB09548EA
| Quantity | 10g |
|---|
Chemical Identifiers
| 13372-81-1 | |
| 147.18 | |
| RUQDOYIAKHIMAN-DAAQNPAKSA-N | |
| 9561350 | |
| O\N=C\C=C\C1=CC=CC=C1 |
| C9H9NO | |
| MFCD00019969 | |
| cinnamaldehyde oxime, 3-phenylacrylaldehyde oxime, phenylacrylaldehyde oxime, e-n-2e-3-phenylprop-2-en-1-ylidene hydroxylamine, n-hydroxy-3-phenylprop-2-en-1-imine, n-3-phenylprop-2-en-1-ylidene hydroxylamine, 3-phenylpropenal oxime, benzeneacrylaldehydeoxime, 3-phenyl-2-propenal oxime, cinnamaldoxime, e + z | |
| (NE)-N-[(E)-3-phenylprop-2-enylidene]hydroxylamine |
Specifications
| 3-Phenylacrylaldehyde oxime | |
| 97% | |
| MFCD00019969 | |
| cinnamaldehyde oxime, 3-phenylacrylaldehyde oxime, phenylacrylaldehyde oxime, e-n-2e-3-phenylprop-2-en-1-ylidene hydroxylamine, n-hydroxy-3-phenylprop-2-en-1-imine, n-3-phenylprop-2-en-1-ylidene hydroxylamine, 3-phenylpropenal oxime, benzeneacrylaldehydeoxime, 3-phenyl-2-propenal oxime, cinnamaldoxime, e + z | |
| O\N=C\C=C\C1=CC=CC=C1 | |
| 147.18 | |
| 147.18 |
| 13372-81-1 | |
| C9H9NO | |
| 10g | |
| RUQDOYIAKHIMAN-DAAQNPAKSA-N | |
| (NE)-N-[(E)-3-phenylprop-2-enylidene]hydroxylamine | |
| 9561350 |