missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylacrylaldehyde oxime, 97%, Thermo Scientific™

Catalog No. p-4812974
Change view
Click to view available options
Quantity:
1g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB09548DA 1g
BTB09548EA 10g
BTB09548EB 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BTB09548DA Supplier Thermo Scientific Chemicals Supplier No. BTB09548DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13372-81-1
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00019969
InChI Key RUQDOYIAKHIMAN-DAAQNPAKSA-N
Synonym cinnamaldehyde oxime, 3-phenylacrylaldehyde oxime, phenylacrylaldehyde oxime, e-n-2e-3-phenylprop-2-en-1-ylidene hydroxylamine, n-hydroxy-3-phenylprop-2-en-1-imine, n-3-phenylprop-2-en-1-ylidene hydroxylamine, 3-phenylpropenal oxime, benzeneacrylaldehydeoxime, 3-phenyl-2-propenal oxime, cinnamaldoxime, e + z
PubChem CID 9561350
IUPAC Name (NE)-N-[(E)-3-phenylprop-2-enylidene]hydroxylamine
SMILES O\N=C\C=C\C1=CC=CC=C1

Specifications

Assay Percent Range 97%
Quantity 1g
Formula Weight 147.18
Chemical Name or Material 3-Phenylacrylaldehyde oxime