missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Phenyl-3-(trifluoromethyl)-3H-diazirine 98.0+%, TCI America™
$305.70 - $1066.24
Chemical Identifiers
| CAS | 73899-14-6 |
|---|---|
| Molecular Formula | C8H5F3N2 |
| Molecular Weight (g/mol) | 186.137 |
| InChI Key | IXEVBKYJNFWMPR-UHFFFAOYSA-N |
| PubChem CID | 11116743 |
| IUPAC Name | 3-phenyl-3-(trifluoromethyl)diazirine |
| SMILES | C1=CC=C(C=C1)C2(N=N2)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P1963200MG
|
TCI America
P1963200MG |
200 mg |
Each for $305.70
|
|
|||||
|
P19631G
|
TCI America
P19631G |
1 g |
Each for $1,066.24
|
|
|||||
Chemical Identifiers
| 73899-14-6 | |
| 186.137 | |
| 11116743 | |
| C1=CC=C(C=C1)C2(N=N2)C(F)(F)F |
| C8H5F3N2 | |
| IXEVBKYJNFWMPR-UHFFFAOYSA-N | |
| 3-phenyl-3-(trifluoromethyl)diazirine |
Specifications
| 73899-14-6 | |
| C8H5F3N2 | |
| IXEVBKYJNFWMPR-UHFFFAOYSA-N | |
| 3-phenyl-3-(trifluoromethyl)diazirine | |
| 11116743 | |
| ≥98.0% (HPLC) | |
| 3-Phenyl-3-(trifluoromethyl)-3H-diazirine |
| Yellow | |
| 200 mg | |
| C1=CC=C(C=C1)C2(N=N2)C(F)(F)F | |
| 186.137 | |
| 186.14 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator