accessibility menu, dialog, popup

UserName

NNFFF

CAS RN 73899-14-6

IUPAC Name: 3-phenyl-3-(trifluoromethyl)-3H-diazirine
Synonyms: 3-phenyl-3-(trifluoromethyl)diazirine
Molecular Weight (g/mol): 186.14
Molecular Formula: C8H5F3N2
InChi Key: IXEVBKYJNFWMPR-UHFFFAOYSA-N
SMILES: FC(F)(F)C1(N=N1)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results