missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-3-(trifluoromethyl)-3H-diazirine 98.0+%, TCI America™

Catalog No. P1963200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1963200MG 200 mg
P19631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1963200MG Supplier TCI America Supplier No. P1963200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 73899-14-6
Molecular Formula C8H5F3N2
Molecular Weight (g/mol) 186.137
InChI Key IXEVBKYJNFWMPR-UHFFFAOYSA-N
PubChem CID 11116743
IUPAC Name 3-phenyl-3-(trifluoromethyl)diazirine
SMILES C1=CC=C(C=C1)C2(N=N2)C(F)(F)F

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 186.14
Percent Purity ≥98.0% (HPLC)
Physical Form Liquid
Chemical Name or Material 3-Phenyl-3-(trifluoromethyl)-3H-diazirine
TSCA No
Recommended Storage Refrigerator