missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-2-propyn-1-ol, 98+%

Catalog No. p-7059019
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4377804 2 g
AA4377809 10 g
AA4377818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4377804 Supplier Thermo Scientific Chemicals Supplier No. 04377804

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1504-58-1 | C9H8O | 132.162 g/mol

3-Phenyl-2-propyn-1-ol is an phenyl acetylene derivative with inhibitory effect on nitrification in soil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1504-58-1
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.162
MDL Number MFCD00040914
InChI Key NITUNGCLDSFVDL-UHFFFAOYSA-N
Synonym 3-phenyl-2-propyn-1-ol, 2-propyn-1-ol, 3-phenyl, 3-phenyl-prop-2-yn-1-ol, 1-hydroxymethyl-2-phenylacetylene, 1-phenyl-1-propyn-3-ol, pubchem13580, 3-phenyl-2-propyn1-ol, 3-phenylpropargyl alcohol, acmc-1bs1u, 1-phenyl-propyn-3-ol
PubChem CID 123115
IUPAC Name 3-phenylprop-2-yn-1-ol
SMILES C1=CC=C(C=C1)C#CCO

Specifications

Density 1.078 g/mL
Boiling Point 129°C to 130°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5867
Quantity 2 g
Beilstein 2040911
Solubility Information Soluble in ether,acetone,benzene,chloroform,ethyl Acetate,methanol.
Formula Weight 132.16
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 3-Phenyl-2-propyn-1-ol
Show More Show Less
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only