missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-2,4-pentanedione 98.0+%, TCI America™

Catalog No. p-7135063
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12971G 1 g
P12975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12971G Supplier TCI America Supplier No. P12971G

Chemical Identifiers

CAS 5910-25-8
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00048223
InChI Key YIWTXSVNRCWBAC-UHFFFAOYSA-N
Synonym 3-Phenylacetylacetone
PubChem CID 233402
IUPAC Name 3-phenylpentane-2,4-dione
SMILES CC(=O)C(C1=CC=CC=C1)C(=O)C

Specifications

Melting Point 56°C
Color White-Yellow
Boiling Point 134°C
Quantity 1 g
Formula Weight 176.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Phenyl-2,4-pentanedione