missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-1,2,4-triazole-5-thiol hydrate, 98%

Catalog No. AAA1242703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1242703 1 g
AAA1242706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1242703 Supplier Thermo Scientific Chemicals Supplier No. A1242703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3414-94-6 | C8H7N3S | 177.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3414-94-6
Molecular Formula C8H7N3S
Molecular Weight (g/mol) 177.23
MDL Number MFCD00051814
InChI Key JRLMMJNORORYPO-UHFFFAOYSA-N
Synonym 5-phenyl-4h-1,2,4-triazole-3-thiol, 5-phenyl-1h-1,2,4-triazole-3-thiol, 3-phenyl-1h-1,2,4-triazole-5-thiol, 3-phenyl-1,2,4-triazole-5-thiol, 5-phenyl-s-triazole-3-thiol, 5-phenyl-4h-1,2,4-triazol-3-yl hydrosulfide, 1h-1,2,4-triazole-3-thiol, 5-phenyl, 1,4-dihydro-5-phenyl-3h-1,2,4-triazole-3-thione, 2-fenil-5-mercapto-1,3,4-triazolo italian, 3h-1,2,4-triazole-3-thione, 1,4-dihydro-5-phenyl
PubChem CID 698268
IUPAC Name 5-phenyl-1,2-dihydro-1,2,4-triazole-3-thione
SMILES S=C1NNC(=N1)C1=CC=CC=C1

Specifications

Melting Point 253°C to 258°C
Quantity 1 g
Beilstein 135556
Formula Weight 177.23 (Anhydrous)
Percent Purity 98%
Chemical Name or Material 3-Phenyl-1,2,4-triazole-5-thiol hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber XZ5351000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only