CAS RN 3414-94-6
IUPAC Name:
5-phenyl-2,3-dihydro-1H-1,2,4-triazole-3-thione
Synonyms:
5-phenyl-1,2-dihydro-1,2,4-triazole-3-thione
Molecular Weight (g/mol):
177.23
Molecular Formula:
C8H7N3S
InChi Key:
JRLMMJNORORYPO-UHFFFAOYSA-N
SMILES:
S=C1NNC(=N1)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
3-Phenyl-1,2,4-triazole-5-thiol hydrate, 98%
3-Phenyl-1,2,4-triazole-5-thiol hydrate, 98%
CAS: 3414-94-6 Molecular Formula: C8H7N3S Molecular Weight (g/mol): 177.23 MDL Number: MFCD00051814 InChI Key: JRLMMJNORORYPO-UHFFFAOYSA-N Synonym: 5-phenyl-4h-1,2,4-triazole-3-thiol,5-phenyl-1h-1,2,4-triazole-3-thiol,3-phenyl-1h-1,2,4-triazole-5-thiol,3-phenyl-1,2,4-triazole-5-thiol,5-phenyl-s-triazole-3-thiol,5-phenyl-4h-1,2,4-triazol-3-yl hydrosulfide,1h-1,2,4-triazole-3-thiol, 5-phenyl,1,4-dihydro-5-phenyl-3h-1,2,4-triazole-3-thione,2-fenil-5-mercapto-1,3,4-triazolo italian,3h-1,2,4-triazole-3-thione, 1,4-dihydro-5-phenyl PubChem CID: 698268 IUPAC Name: 5-phenyl-1,2-dihydro-1,2,4-triazole-3-thione SMILES: S=C1NNC(=N1)C1=CC=CC=C1