missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxytoluene 98.0+%, TCI America™

Catalog No. p-7134797
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P083125ML 25 mL
P0831100ML 100 mL
P0831500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P083125ML Supplier TCI America Supplier No. P083125ML
Only null left

Chemical Identifiers

CAS 3586-14-9
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.24
MDL Number MFCD00008531
InChI Key UDONPJKEOAWFGI-UHFFFAOYSA-N
Synonym 3-phenoxytoluene, 3-methyldiphenyl ether, m-phenoxytoluene, phenyl m-tolyl ether, ether, phenyl m-tolyl, benzene, 1-methyl-3-phenoxy, m-methylphenyl phenyl ether, 3-methylphenyl phenyl ether, unii-2zzi1z67wr
PubChem CID 19165
IUPAC Name 1-methyl-3-phenoxybenzene
SMILES CC1=CC=CC(OC2=CC=CC=C2)=C1

Specifications

Color Colorless
Boiling Point 272°C
Quantity 25 mL
UN Number 3082
Formula Weight 184.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Phenoxytoluene
EINECSNumber (3)-0752
RTECSNumber DA6142000
TSCA Yes