missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxytoluene, 97%

Catalog No. p-7026095
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2189914 25 g
AAB2189922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2189914 Supplier Thermo Scientific Chemicals Supplier No. B2189914

CAS: 3586-14-9 | C13H12O | 184.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3586-14-9
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.24
MDL Number MFCD00008531
InChI Key UDONPJKEOAWFGI-UHFFFAOYSA-N
Synonym 3-phenoxytoluene, 3-methyldiphenyl ether, m-phenoxytoluene, phenyl m-tolyl ether, ether, phenyl m-tolyl, benzene, 1-methyl-3-phenoxy, m-methylphenyl phenyl ether, 3-methylphenyl phenyl ether, unii-2zzi1z67wr
PubChem CID 19165
IUPAC Name 1-methyl-3-phenoxybenzene
SMILES CC1=CC=CC(OC2=CC=CC=C2)=C1

Specifications

Density 1.052
Boiling Point 271°C to 273°C
Flash Point >110°C (230°F)
Refractive Index 1.574
Quantity 25 g
Beilstein 2045714
Formula Weight 184.24
Percent Purity 97%
Chemical Name or Material 3-Phenoxytoluene
EINECSNumber 222-716-4
RTECSNumber DA6142000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only