missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxyphenylacetic acid, 98%

Catalog No. p-7045938
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1252403 1 g
AAL1252406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1252403 Supplier Thermo Scientific Chemicals Supplier No. L1252403
Only null left

CAS: 32852-81-6 | C14H12O3 | 228.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32852-81-6
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.247
MDL Number MFCD00016826
InChI Key LEMRHTTWKDVQEI-UHFFFAOYSA-N
Synonym 3-phenoxyphenylacetic acid, 2-3-phenoxyphenyl acetic acid, 3-phenoxyphenyl acetic acid, 3-phenoxy-phenyl-acetic acid, benzeneacetic acid, 3-phenoxy, 3-phenoxyphenylaceticacid, m-phenoxyphenylacetic acid, acmc-20a35b, 3-phenoxybenzeneacetic acid, 3-phenoxyphenyl-acetic acid
PubChem CID 141749
IUPAC Name 2-(3-phenoxyphenyl)acetic acid
SMILES C1=CC=C(C=C1)OC2=CC=CC(=C2)CC(=O)O

Specifications

Melting Point 88°C to 90°C
Quantity 1 g
Beilstein 2730540
Formula Weight 228.25
Percent Purity 98%
Chemical Name or Material 3-Phenoxyphenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only