missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Phenoxymethyl)benzoic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC63601DA
Specifications
| 3-(phenoxymethyl)benzoic acid | |
| 97% | |
| C14H12O3 | |
| 3-phenoxymethyl benzoic acid, benzoic acid,3-phenoxymethyl, 4rg | |
| C1=CC=C(C=C1)OCC2=CC=CC(=C2)C(=O)O | |
| 228.247 | |
| 228.25 |
| 31719-75-2 | |
| Amber Glass Bottle | |
| 1g | |
| URLYREZIPSJJQU-UHFFFAOYSA-N | |
| 3-(phenoxymethyl)benzoic acid | |
| 3729884 | |
| 97% |
Chemical Identifiers
| 31719-75-2 | |
| 228.247 | |
| 3-phenoxymethyl benzoic acid, benzoic acid,3-phenoxymethyl, 4rg | |
| 3-(phenoxymethyl)benzoic acid |
| C14H12O3 | |
| URLYREZIPSJJQU-UHFFFAOYSA-N | |
| 3729884 | |
| C1=CC=C(C=C1)OCC2=CC=CC(=C2)C(=O)O |
Safety and Handling
GHS Signal Word: Warning