missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxybenzoic Acid 98.0+%, TCI America™

Supplier:  TCI America P125325G

 View more versions of this product

Catalog No. P125325G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Phenoxybenzoic Acid
147°C
C13H10O3
25 g
NXTDJHZGHOFSQG-UHFFFAOYSA-N
3-phenoxybenzoic acid
19539
214.22
Crystalline Powder
3739-38-6
White
MFCD00002498
m-phenoxybenzoic acid, benzoic acid, 3-phenoxy, 3-carboxydiphenyl ether, 3-phenoxybenzoicacid, unii-69dc2655vh, benzoic acid, m-phenoxy, 3-phenoxy-benzoic acid, chembl663, m-phenoxybenzoic acid for cis-isomer, vjj
C1=CC=C(C=C1)OC2=CC=CC(=C2)C(=O)O
214.22
CHEBI:72631
≥98.0% (GC,T)

Chemical Identifiers

3739-38-6
214.22
NXTDJHZGHOFSQG-UHFFFAOYSA-N
19539
3-phenoxybenzoic acid
C13H10O3
MFCD00002498
m-phenoxybenzoic acid, benzoic acid, 3-phenoxy, 3-carboxydiphenyl ether, 3-phenoxybenzoicacid, unii-69dc2655vh, benzoic acid, m-phenoxy, 3-phenoxy-benzoic acid, chembl663, m-phenoxybenzoic acid for cis-isomer, vjj
CHEBI:72631
C1=CC=C(C=C1)OC2=CC=CC(=C2)C(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0995

RTECSNumber : DH6293500

TSCA : No

SDS
Documents

Documents

Promotions

Promotions