missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxybenzoic Acid 98.0+%, TCI America™

Catalog No. P12535G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12535G 5 g
P125325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12535G Supplier TCI America Supplier No. P12535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3739-38-6
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00002498
InChI Key NXTDJHZGHOFSQG-UHFFFAOYSA-N
Synonym m-phenoxybenzoic acid, benzoic acid, 3-phenoxy, 3-carboxydiphenyl ether, 3-phenoxybenzoicacid, unii-69dc2655vh, benzoic acid, m-phenoxy, 3-phenoxy-benzoic acid, chembl663, m-phenoxybenzoic acid for cis-isomer, vjj
PubChem CID 19539
ChEBI CHEBI:72631
IUPAC Name 3-phenoxybenzoic acid
SMILES C1=CC=C(C=C1)OC2=CC=CC(=C2)C(=O)O

Specifications

Melting Point 147°C
Color White
Quantity 5 g
Formula Weight 214.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Phenoxybenzoic Acid
EINECSNumber (3)-0995
RTECSNumber DH6293500
TSCA No