missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxyanisole, 97%

Catalog No. AAH3248206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3248206 5 g
AAH3248214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3248206 Supplier Thermo Scientific Chemicals Supplier No. H3248206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1655-68-1 | C13H12O2 | 200.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

Chemical Identifiers

CAS 1655-68-1
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.24
MDL Number MFCD00455605
InChI Key CBVXNDCIOLXDFD-UHFFFAOYSA-N
PubChem CID 74252
IUPAC Name 1-methoxy-3-phenoxybenzene
SMILES COC1=CC=CC(OC2=CC=CC=C2)=C1

Specifications

Odor Odorless
Refractive Index 1.579
Quantity 5 g
Synonym benzene, 1-methoxy-3-phenoxy, m-methoxyphenyl phenyl ether, m-phenoxyanisole, 3-phenoxyanisole, m-phenoxymethoxybenzene, 3-methoxyphenoxybenzene, m-phenoxyphenol monomethyl ether, 3-methoxydiphenyl ether, benzene,1-methoxy-3-phenoxy, benzene, 1-methoxy-3-phenyloxy
Solubility Information Insoluble in water.
Formula Weight 200.23
Percent Purity 97%
Chemical Name or Material 3-Phenoxyanisole
EINECSNumber 216-748-8
RTECSNumber DA5350200
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only