missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxyaniline, 98%

Catalog No. AAA1749506
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1749506 5 g
AAA1749514 25 g
AAA1749522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1749506 Supplier Thermo Scientific Chemicals Supplier No. A1749506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3586-12-7 | C12H11NO | 185.23 g/mol

3-Phenoxyaniline was used in the preparation of 4-phenoxyaniline-3?-bromopropionamide. It is also used in the design, synthesis and evaluation of potential inhibitors of HIV gp120-CD4 interactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Phenoxyaniline was used in the preparation of 4-phenoxyaniline-3′-bromopropionamide. It is also used in the design, synthesis and evaluation of potential inhibitors of HIV gp120-CD4 interactions.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 3586-12-7
MDL Number MFCD00041891

Specifications

Melting Point 35°C to 37°C
Density 1.1583
Boiling Point 329°C to 330°C
Flash Point >110°C (230°F)
Molecular Formula C6H10O3
Refractive Index 1.1583
Quantity 5 g
Synonym neopentylene carbonate, 2,2-dimethyltrimethylene carbonate, 5,5-dimethyl-1,3-diox-2-one, acmc-1adx1, 3-nitro-5-chlorobenzenesulfonamide, 5,5-dimethyltrimethylene carbonate, 5,5-dimethyl-1,3-dioxane-2-one, 5,5-dimethyl-2-oxo-1,3-dioxane, 1,3-dioxan-2-one,5,5-dimethyl, 5,5-dimethyl-1,3 dioxan-2-one
Solubility Information Slightly soluble in water.
InChI Key JRFXQKZEGILCCO-UHFFFAOYSA-N
SMILES CC1(COC(=O)OC1)C
IUPAC Name 5,5-dimethyl-1,3-dioxan-2-one
Molecular Weight (g/mol) 130.14
PubChem CID 137984
Formula Weight 185.23
Percent Purity 98%
Chemical Name or Material 3-Phenoxyaniline
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 222-715-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only