missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Phenoxy-1-propanol 98.0+%, TCI America™
$67.38 - $180.97
Chemical Identifiers
| CAS | 6180-61-6 |
|---|---|
| Molecular Formula | C9H12O2 |
| Molecular Weight (g/mol) | 152.19 |
| MDL Number | MFCD00039549 |
| InChI Key | AWVDYRFLCAZENH-UHFFFAOYSA-N |
| Synonym | 3-phenoxy-1-propanol, 1-propanol, 3-phenoxy, 3-phenoxy 1-propanol, unii-515pbp1u72, propoxylated phenol, ph 4p, polypropylene glycol phenyl ether, 3-phenoxypropanol, 3-phenyloxypropanol, polypropylene glycol monophenyl ether |
| PubChem CID | 80319 |
| IUPAC Name | 3-phenoxypropan-1-ol |
| SMILES | OCCCOC1=CC=CC=C1 |
Chemical Identifiers
| 6180-61-6 | |
| 152.19 | |
| AWVDYRFLCAZENH-UHFFFAOYSA-N | |
| 80319 | |
| OCCCOC1=CC=CC=C1 |
| C9H12O2 | |
| MFCD00039549 | |
| 3-phenoxy-1-propanol, 1-propanol, 3-phenoxy, 3-phenoxy 1-propanol, unii-515pbp1u72, propoxylated phenol, ph 4p, polypropylene glycol phenyl ether, 3-phenoxypropanol, 3-phenyloxypropanol, polypropylene glycol monophenyl ether | |
| 3-phenoxypropan-1-ol |
Specifications
| 6180-61-6 | |
| 153°C | |
| MFCD00039549 | |
| 3-phenoxy-1-propanol, 1-propanol, 3-phenoxy, 3-phenoxy 1-propanol, unii-515pbp1u72, propoxylated phenol, ph 4p, polypropylene glycol phenyl ether, 3-phenoxypropanol, 3-phenyloxypropanol, polypropylene glycol monophenyl ether | |
| OCCCOC1=CC=CC=C1 | |
| 152.19 | |
| 152.19 | |
| Liquid |
| Colorless | |
| C9H12O2 | |
| 5 g | |
| AWVDYRFLCAZENH-UHFFFAOYSA-N | |
| 3-phenoxypropan-1-ol | |
| 80319 | |
| ≥98.0% (GC) | |
| 3-Phenoxy-1-propanol |
Safety and Handling
TSCA : No