missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxy-1-propanol, 97%

CAS: 6180-61-6 | C9H12O2 | 152.19 g/mol

$284.44 - $284.44

Chemical Identifiers

CAS 6180-61-6
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00039549
InChI Key AWVDYRFLCAZENH-UHFFFAOYSA-N
Synonym 3-phenoxy-1-propanol, 1-propanol, 3-phenoxy, 3-phenoxy 1-propanol, unii-515pbp1u72, propoxylated phenol, ph 4p, polypropylene glycol phenyl ether, 3-phenoxypropanol, 3-phenyloxypropanol, polypropylene glycol monophenyl ether
PubChem CID 80319
IUPAC Name 3-phenoxypropan-1-ol
SMILES OCCCOC1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC186980250
SDS View Documents
Thermo Scientific Chemicals
186980250
25 g Glass bottle
Each for $284.44
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6180-61-6
152.19
AWVDYRFLCAZENH-UHFFFAOYSA-N
80319
OCCCOC1=CC=CC=C1
C9H12O2
MFCD00039549
3-phenoxy-1-propanol, 1-propanol, 3-phenoxy, 3-phenoxy 1-propanol, unii-515pbp1u72, propoxylated phenol, ph 4p, polypropylene glycol phenyl ether, 3-phenoxypropanol, 3-phenyloxypropanol, polypropylene glycol monophenyl ether
3-phenoxypropan-1-ol

Specifications

6180-61-6
100.0
1.0400g/mL
>112°C
97%
C9H12O2
C6H5O(CH2)3OH
25 g
1.04
AWVDYRFLCAZENH-UHFFFAOYSA-N
3-phenoxypropan-1-ol
80319
97%
3-Phenoxy-1-propanol, 97%
96.0
Colorless
83°C to 85°C
Authentic
Glass bottle
1.5280 to 1.53
MFCD00039549
06, 147
3-phenoxy-1-propanol, 1-propanol, 3-phenoxy, 3-phenoxy 1-propanol, unii-515pbp1u72, propoxylated phenol, ph 4p, polypropylene glycol phenyl ether, 3-phenoxypropanol, 3-phenyloxypropanol, polypropylene glycol monophenyl ether
OCCCOC1=CC=CC=C1
152.19
152.19
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 228-228-8

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only