missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Pentanone, 99%
Description
3-Pentanone is mainly used as a solvent in paint and a precursor to vitamin E. It is used as a reagent to synthesize ethyl 2-cyano-3,3-diethylacrylate by Knoevenagel condensation. It also shows anticonvulsant effect in several types of mouse seizure models. It serves as an intermediate in the manufacture of pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Pentanone is mainly used as a solvent in paint and a precursor to vitamin E. It is used as a reagent to synthesize ethyl 2-cyano-3,3-diethylacrylate by Knoevenagel condensation. It also shows anticonvulsant effect in several types of mouse seizure models. It serves as an intermediate in the manufacture of pharmaceuticals.
Solubility
Soluble in water, alcohol, ether and carbonterachloride. Slightly miscible with ethanol and acetone.
Notes
Incompatible with strong oxidizing agents, reducing agents and strong bases.
3-Pentanone is mainly used as a solvent in paint and a precursor to vitamin E. It is used as a reagent to synthesize ethyl 2-cyano-3,3-diethylacrylate by Knoevenagel condensation. It also shows anticonvulsant effect in several types of mouse seizure models. It serves as an intermediate in the manufacture of pharmaceuticals.
Solubility
Soluble in water, alcohol, ether and carbonterachloride. Slightly miscible with ethanol and acetone.
Notes
Incompatible with strong oxidizing agents, reducing agents and strong bases.
Specifications
Specifications
| Melting Point | -39°C |
| Density | 0.812 |
| Boiling Point | 102°C to 103°C |
| Flash Point | 7°C (45°F) |
| Odor | Acetone-like |
| Refractive Index | 1.392 |
| Quantity | 100 mL |
| UN Number | UN1156 |
| Beilstein | 635749 |
| Merck Index | 14,3121 |
| Show More |
Safety and Handling
| Hazard Category | H225-H319-H335-H336 |
| Hazard Statement | GHS H Statement H225-H335-H336 Highly flammable liquid and vapour. May cause respiratory irritation. May cause drowsiness or dizziness. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIETHYL KETONE |
| EINECSNumber | 202-490-3 |
| RTECSNumber | SA8050000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only