accessibility menu, dialog, popup

UserName

CH3OH3C

CAS RN 96-22-0

IUPAC Name: pentan-3-one
Synonyms: 3-pentanone
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: FDPIMTJIUBPUKL-UHFFFAOYSA-N
SMILES: CCC(=O)CC
Boiling Point 
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (2)