missing translation for 'onlineSavingsMsg'
Learn More

3-Pentanone, 99%

Catalog No. AAA15297AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA15297AE 100 mL
AAA15297AP 500 mL
AAA152970F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA15297AE Supplier Thermo Scientific Chemicals Supplier No. A15297AE

CAS: 96-22-0 | C5H10O | 86.13 g/mol

3-Pentanone is mainly used as a solvent in paint and a precursor to vitamin E. It is used as a reagent to synthesize ethyl 2-cyano-3,3-diethylacrylate by Knoevenagel condensation. It also shows anticonvulsant effect in several types of mouse seizure models. It serves as an intermediate in the manufacture of pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Pentanone is mainly used as a solvent in paint and a precursor to vitamin E. It is used as a reagent to synthesize ethyl 2-cyano-3,3-diethylacrylate by Knoevenagel condensation. It also shows anticonvulsant effect in several types of mouse seizure models. It serves as an intermediate in the manufacture of pharmaceuticals.

Solubility
Soluble in water, alcohol, ether and carbonterachloride. Slightly miscible with ethanol and acetone.

Notes
Incompatible with strong oxidizing agents, reducing agents and strong bases.

Chemical Identifiers

CAS 96-22-0
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00009320
InChI Key FDPIMTJIUBPUKL-UHFFFAOYSA-N
Synonym 3-pentanone, diethyl ketone, propione, ethyl ketone, dimethylacetone, metacetone, methacetone, diethylcetone, ethyl propionyl, diethylketone
PubChem CID 7288
ChEBI CHEBI:67886
IUPAC Name pentan-3-one
SMILES CCC(=O)CC

Specifications

Melting Point -39°C
Density 0.812
Boiling Point 102°C to 103°C
Flash Point 7°C (45°F)
Odor Acetone-like
Refractive Index 1.392
Quantity 100 mL
UN Number UN1156
Beilstein 635749
Merck Index 14,3121
Solubility Information Soluble in water,alcohol,ether and carbonterachloride. Slightly miscible with ethanol and acetone.
Formula Weight 86.13
Percent Purity 99%
Chemical Name or Material 3-Pentanone
Show More Show Less
Hazard Category H225-H319-H335-H336
Hazard Statement GHS H Statement
H225-H335-H336
Highly flammable liquid and vapour.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIETHYL KETONE
EINECSNumber 202-490-3
RTECSNumber SA8050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only