Learn More
3-(Pentafluorothio)aniline, 97%
CAS: 2993-22-8 | C6H6F5NS | 219.17 g/mol
$132.92 - $430.74
Chemical Identifiers
| CAS | 2993-22-8 |
|---|---|
| Molecular Formula | C6H6F5NS |
| Molecular Weight (g/mol) | 219.17 |
| MDL Number | MFCD00674199 |
| InChI Key | NUFLICUHOXHWER-UHFFFAOYSA-N |
| Synonym | 3-aminophenylsulfur pentafluoride, 3-pentafluorothio aniline, 3-pentafluorosulfanyl aniline, 3-aminophenylsulphur pentafluoride, sulfur, 3-aminophenyl pentafluoro-, oc-6-21-9ci, 3-pentafluoro-??-sulfanyl aniline, acmc-1cou4, 3-pentafluorosulfanylaniline, 3-pentafluorosulphanylaniline, 3-pentafluorosulfur aniline |
| PubChem CID | 2779188 |
| IUPAC Name | 3-(pentafluoro-$l^{6}-sulfanyl)aniline |
| SMILES | NC1=CC(=CC=C1)S(F)(F)(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3519003
|
Thermo Scientific Chemicals
H3519003 |
1 g |
N/A
|
|
|||||
|
AAH3519006
|
Thermo Scientific Chemicals
H3519006 |
5 g |
N/A
|
|
|||||
Description
Reduction of pentafluoroaniline (PFA) leads to rapid fluoride elimination to form the aminotetrafluorophenyl radical. Homolytic reactions of perfluoroaromatic compounds(Formation of pentafluorophenyl radicals from pentafluoroaniline, and their reactions with aromatic compounds).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReduction of pentafluoroaniline (PFA) leads to rapid fluoride elimination to form the aminotetrafluorophenyl radical. Homolytic reactions of perfluoroaromatic compounds(Formation of pentafluorophenyl radicals from pentafluoroaniline, and their reactions with aromatic compounds).
Solubility
Soluble in methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store in a refrigerator. Store under inert gas.
Chemical Identifiers
| 2993-22-8 | |
| 219.17 | |
| NUFLICUHOXHWER-UHFFFAOYSA-N | |
| 2779188 | |
| NC1=CC(=CC=C1)S(F)(F)(F)(F)F |
| C6H6F5NS | |
| MFCD00674199 | |
| 3-aminophenylsulfur pentafluoride, 3-pentafluorothio aniline, 3-pentafluorosulfanyl aniline, 3-aminophenylsulphur pentafluoride, sulfur, 3-aminophenyl pentafluoro-, oc-6-21-9ci, 3-pentafluoro-??-sulfanyl aniline, acmc-1cou4, 3-pentafluorosulfanylaniline, 3-pentafluorosulphanylaniline, 3-pentafluorosulfur aniline | |
| 3-(pentafluoro-$l^{6}-sulfanyl)aniline |
Specifications
| 2993-22-8 | |
| MFCD00674199 | |
| UN2811 | |
| Soluble in methanol. | |
| NC1=CC(=CC=C1)S(F)(F)(F)(F)F | |
| 219.17 | |
| 218.17 | |
| 3-(Pentafluorothio)aniline |
| C6H6F5NS | |
| 1 g | |
| 3-aminophenylsulfur pentafluoride, 3-pentafluorothio aniline, 3-pentafluorosulfanyl aniline, 3-aminophenylsulphur pentafluoride, sulfur, 3-aminophenyl pentafluoro-, oc-6-21-9ci, 3-pentafluoro-??-sulfanyl aniline, acmc-1cou4, 3-pentafluorosulfanylaniline, 3-pentafluorosulphanylaniline, 3-pentafluorosulfur aniline | |
| NUFLICUHOXHWER-UHFFFAOYSA-N | |
| 3-(pentafluoro-$l^{6}-sulfanyl)aniline | |
| 2779188 | |
| 97% |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only