accessibility menu, dialog, popup

UserName

SFFFFFH2N

CAS RN 2993-22-8

IUPAC Name: 3-(pentafluoro-λ⁶-sulfanyl)aniline
Synonyms: 3-(pentafluoro-λ⁶-sulfanyl)aniline
Molecular Weight (g/mol): 219.17
Molecular Formula: C6H6F5NS
InChi Key: NUFLICUHOXHWER-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results