missing translation for 'onlineSavingsMsg'
Learn More

3-(Pentafluorothio)aniline, 97%

Catalog No. AAH3519006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3519006 5 g
AAH3519003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3519006 Supplier Thermo Scientific Chemicals Supplier No. H3519006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 2993-22-8 | C6H6F5NS | 219.17 g/mol

Reduction of pentafluoroaniline (PFA) leads to rapid fluoride elimination to form the aminotetrafluorophenyl radical. Homolytic reactions of perfluoroaromatic compounds(Formation of pentafluorophenyl radicals from pentafluoroaniline, and their reactions with aromatic compounds).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reduction of pentafluoroaniline (PFA) leads to rapid fluoride elimination to form the aminotetrafluorophenyl radical. Homolytic reactions of perfluoroaromatic compounds(Formation of pentafluorophenyl radicals from pentafluoroaniline, and their reactions with aromatic compounds).

Solubility
Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store in a refrigerator. Store under inert gas.

Chemical Identifiers

CAS 2993-22-8
Molecular Formula C6H6F5NS
Molecular Weight (g/mol) 219.17
MDL Number MFCD00674199
InChI Key NUFLICUHOXHWER-UHFFFAOYSA-N
Synonym 3-aminophenylsulfur pentafluoride, 3-pentafluorothio aniline, 3-pentafluorosulfanyl aniline, 3-aminophenylsulphur pentafluoride, sulfur, 3-aminophenyl pentafluoro-, oc-6-21-9ci, 3-pentafluoro-??-sulfanyl aniline, acmc-1cou4, 3-pentafluorosulfanylaniline, 3-pentafluorosulphanylaniline, 3-pentafluorosulfur aniline
PubChem CID 2779188
IUPAC Name 3-(pentafluoro-$l^{6}-sulfanyl)aniline
SMILES NC1=CC(=CC=C1)S(F)(F)(F)(F)F

Specifications

Quantity 5 g
UN Number UN2811
Solubility Information Soluble in methanol.
Formula Weight 218.17
Percent Purity 97%
Chemical Name or Material 3-(Pentafluorothio)aniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only