missing translation for 'onlineSavingsMsg'
Learn More

3-Oxobutanenitrile, 96%

Catalog No. AAH64536MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64536MD 250 mg
AAH6453603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH64536MD Supplier Thermo Scientific Chemicals Supplier No. H64536MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2469-99-0 | C4H5NO | 83.09 g/mol

It is used as pharmaceutical raw material and intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical raw material and intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 2469-99-0
Molecular Formula C4H5NO
Molecular Weight (g/mol) 83.09
MDL Number MFCD05861381
InChI Key OPXYNEYEDHAXOM-UHFFFAOYSA-N
Synonym 3-oxobutyronitrile, 3-oxo-butyronitrile, butanenitrile, 3-oxo, cyanoacetone, cyanaceton, a-cyanoacetone, acetoacetonitrile, acetylacetonitrile, 1-cyano-2-propanone, alpha-acetylacetonitrile
PubChem CID 75579
IUPAC Name 3-oxobutanenitrile
SMILES CC(=O)CC#N

Specifications

Color Colorless to Yellow
Boiling Point 92°C to 94°C (10 mmHg)
Refractive Index 1.4285
Linear Formula CH3COCH2CN
Quantity 250 mg
UN Number UN3276
Beilstein 1633584
Solubility Information Slightly soluble in water.
Formula Weight 83.09
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3-Oxobutanenitrile
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 219-590-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only