accessibility menu, dialog, popup

UserName

CH3ON

CAS RN 2469-99-0

IUPAC Name: 3-oxobutanenitrile
Synonyms: 3-oxobutanenitrile
Molecular Weight (g/mol): 83.09
Molecular Formula: C4H5NO
InChi Key: OPXYNEYEDHAXOM-UHFFFAOYSA-N
SMILES: CC(=O)CC#N
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)