missing translation for 'onlineSavingsMsg'
Learn More

3-Oxo-N-(4-trifluoromethylphenyl)butyramide 98.0+%, TCI America™

Catalog No. O0444200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0444200MG 200 mg
O04441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O0444200MG Supplier TCI America Supplier No. O0444200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 351-87-1
Molecular Formula C11H10F3NO2
Molecular Weight (g/mol) 245.201
MDL Number MFCD00231301
InChI Key GGDAWUSJZZHVBR-UHFFFAOYSA-N
Synonym 4′C-(Trifluoromethyl)acetoacetanilide
PubChem CID 1472078
IUPAC Name 3-oxo-N-[4-(trifluoromethyl)phenyl]butanamide
SMILES CC(=O)CC(=O)NC1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 154°C
Color White-Yellow
Quantity 200 mg
Formula Weight 245.20
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3-Oxo-N-(4-trifluoromethylphenyl)butyramide