Learn More
3-Oxetanone, 95%
CAS: 6704-31-0 | C3H4O2 | 72.063 g/mol
Supplier: Thermo Scientific Chemicals H5787006
Description
3-Oxetanone is used as a starting material for the preparation of other oxetanes compounds, which are of pharmacological interest. It is also considered as an object for theoretical studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Oxetanone is used as a starting material for the preparation of other oxetanes compounds, which are of pharmacological interest. It is also considered as an object for theoretical studies.
Solubility
Miscible with tetrahydrofuran.
Notes
Incompatible with strong oxidizing agents. Heat sensitive.
Specifications
| 3-Oxetanone | |
| 1.124 | |
| 26°C (79°F) | |
| 1.426 | |
| 5 g | |
| 3-oxetanone, 1,3-epoxy-2-propanone, 3-oxooxetane, oxetane-3-one, oxetan-3-on, 1,3-epoxypropanone, pubchem15880, 3-oxentanone, acmc-209nxj, ksc358e8n | |
| ROADCYAOHVSOLQ-UHFFFAOYSA-N | |
| oxetan-3-one | |
| 15024254 | |
| 95% |
| 6704-31-0 | |
| 140°C | |
| C3H4O2 | |
| MFCD09263255 | |
| UN1993 | |
| Miscible with tetrahydrofuran. | |
| C1C(=O)CO1 | |
| 72.063 | |
| 72.06 |
Chemical Identifiers
| 6704-31-0 | |
| 72.063 | |
| ROADCYAOHVSOLQ-UHFFFAOYSA-N | |
| 15024254 | |
| C1C(=O)CO1 |
| C3H4O2 | |
| MFCD09263255 | |
| 3-oxetanone, 1,3-epoxy-2-propanone, 3-oxooxetane, oxetane-3-one, oxetan-3-on, 1,3-epoxypropanone, pubchem15880, 3-oxentanone, acmc-209nxj, ksc358e8n | |
| oxetan-3-one |
Safety and Handling
GHS H Statement
H301-H318-H226-H315-H317-H335
Toxic if swallowed.
Causes serious eye damage.
Flammable liquid and vapor.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Store at -20°C; Store under Argon
RUO – Research Use Only