missing translation for 'onlineSavingsMsg'
Learn More

3-Oxetanone, 95%

Catalog No. p-7038817
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5787003 1 g
AAH5787006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5787003 Supplier Thermo Scientific Chemicals Supplier No. H5787003
Only null left

CAS: 6704-31-0 | C3H4O2 | 72.063 g/mol

3-Oxetanone is used as a starting material for the preparation of other oxetanes compounds, which are of pharmacological interest. It is also considered as an object for theoretical studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Oxetanone is used as a starting material for the preparation of other oxetanes compounds, which are of pharmacological interest. It is also considered as an object for theoretical studies.

Solubility
Miscible with tetrahydrofuran.

Notes
Incompatible with strong oxidizing agents. Heat sensitive.

Chemical Identifiers

CAS 6704-31-0
Molecular Formula C3H4O2
Molecular Weight (g/mol) 72.063
MDL Number MFCD09263255
InChI Key ROADCYAOHVSOLQ-UHFFFAOYSA-N
Synonym 3-oxetanone, 1,3-epoxy-2-propanone, 3-oxooxetane, oxetane-3-one, oxetan-3-on, 1,3-epoxypropanone, pubchem15880, 3-oxentanone, acmc-209nxj, ksc358e8n
PubChem CID 15024254
IUPAC Name oxetan-3-one
SMILES C1C(=O)CO1

Specifications

Density 1.124
Boiling Point 140°C
Flash Point 26°C (79°F)
Refractive Index 1.426
Quantity 1 g
UN Number UN1993
Solubility Information Miscible with tetrahydrofuran.
Formula Weight 72.06
Percent Purity 95%
Chemical Name or Material 3-Oxetanone
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H301-H318-H226-H315-H317-H335
Toxic if swallowed.
Causes serious eye damage.
Flammable liquid and vapor.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Store at -20°C; Store under Argon

RUO – Research Use Only