missing translation for 'onlineSavingsMsg'
Learn More

3-Oxetanol 96.0+%, TCI America™

Catalog No. O04091G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04091G 1 g
O04095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O04091G Supplier TCI America Supplier No. O04091G

Chemical Identifiers

CAS 7748-36-9
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.08
MDL Number MFCD09056790
InChI Key QMLWSAXEQSBAAQ-UHFFFAOYSA-N
Synonym 3-hydroxyoxetane, 3-oxetanol, 1,3-epoxy-2-propanol, oxetane-3-ol, 3-hydroxy-oxetane, 3-hydroxy oxetane, pubchem17367, ksc380k3t, 1,3-epoxypropan-2-ol
PubChem CID 9942117
IUPAC Name oxetan-3-ol
SMILES OC1COC1

Specifications

Color Yellow
Boiling Point 50°C
Quantity 1 g
Formula Weight 74.08
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Oxetanol
TSCA No
Recommended Storage Refrigerator