missing translation for 'onlineSavingsMsg'
Learn More

3-Oxetanol, 95%

Catalog No. p-7038866
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5792803 1 g
AAH5792806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5792803 Supplier Thermo Scientific Chemicals Supplier No. H5792803
Only null left

CAS: 7748-36-9 | C3H6O2 | 74.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7748-36-9
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.08
MDL Number MFCD09056790
InChI Key QMLWSAXEQSBAAQ-UHFFFAOYSA-N
Synonym 3-hydroxyoxetane, 3-oxetanol, 1,3-epoxy-2-propanol, oxetane-3-ol, 3-hydroxy-oxetane, 3-hydroxy oxetane, pubchem17367, ksc380k3t, 1,3-epoxypropan-2-ol
PubChem CID 9942117
IUPAC Name oxetan-3-ol
SMILES OC1COC1

Specifications

Density 1.167
Boiling Point 153°C
Flash Point 87°C (189°F)
Refractive Index 1.438
Quantity 1 g
Formula Weight 74.08
Percent Purity 95%
Chemical Name or Material 3-Oxetanol
Hazard Category H227-H302-H315-H318-H335
Hazard Statement GHS H Statement
H301-H318-H227-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only