missing translation for 'onlineSavingsMsg'
Learn More

3-Octyn-1-ol, 96%

Catalog No. AAL0541414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0541414 25 g
AAL0541406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0541414 Supplier Thermo Scientific Chemicals Supplier No. L0541414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14916-80-4 | C8H14O | 126.199 g/mol

3-Octyn-1-ol, is used for a study of the reduction of acetylenic compounds, including this alcohol, to (E)-alkenes by alkali metals in liquid ammonia. Some other applications includes as corrosion inhibitor formulation, metal complex solution, solvent stabilizer, biodegradable chemical intermediate to make pharmaceuticals, chemical compound synthesis and reagents

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Octyn-1-ol, is used for a study of the reduction of acetylenic compounds, including this alcohol, to (E)-alkenes by alkali metals in liquid ammonia. Some other applications includes as corrosion inhibitor formulation, metal complex solution, solvent stabilizer, biodegradable chemical intermediate to make pharmaceuticals, chemical compound synthesis and reagents

Solubility
It is soluble in water.

Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 14916-80-4
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00039552
InChI Key LRZGRGVRZSDRTK-UHFFFAOYSA-N
Synonym 3-octyn-1-ol, 3-octyne-1-ol, pubchem13072, acmc-1c9k2, 3-octyn-1-ol 5g
PubChem CID 84694
IUPAC Name oct-3-yn-1-ol
SMILES CCCCC#CCCO

Specifications

Density 0.883
Boiling Point 86°C to 88°C (9 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.454
Quantity 25 g
Beilstein 1739950
Solubility Information It is soluble in water.
Formula Weight 126.2
Percent Purity 96%
Chemical Name or Material 3-Octyn-1-ol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 238-986-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only