accessibility menu, dialog, popup

UserName

CAS RN 14916-80-4

IUPAC Name: oct-3-yn-1-ol
Synonyms: oct-3-yn-1-ol
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: LRZGRGVRZSDRTK-UHFFFAOYSA-N
SMILES: CCCCC#CCCO
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)