missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Octen-2-one 96.0+%, TCI America™
Supplier: TCI America O02525ML
Specifications
| 3-Octen-2-one | |
| Yellow | |
| C8H14O | |
| 5 mL | |
| 3-octen-2-one, trans-3-octen-2-one, e-3-octen-2-one, 3e-3-octen-2-one, e-oct-3-en-2-one, unii-m26ah283xv, 3-octen-2-one, e, 3e-octen-2-one, 3e-oct-3-en-2-one, hex-1-enyl methyl ketone | |
| CCCCC=CC(=O)C | |
| 126.199 | |
| 126.20 | |
| Liquid |
| 1669-44-9 | |
| 78°C | |
| MFCD00015565 | |
| 1224 | |
| ZCFOBLITZWHNNC-VOTSOKGWSA-N | |
| (E)-oct-3-en-2-one | |
| 5363229 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 1669-44-9 | |
| 126.199 | |
| ZCFOBLITZWHNNC-VOTSOKGWSA-N | |
| 5363229 | |
| CCCCC=CC(=O)C |
| C8H14O | |
| MFCD00015565 | |
| 3-octen-2-one, trans-3-octen-2-one, e-3-octen-2-one, 3e-3-octen-2-one, e-oct-3-en-2-one, unii-m26ah283xv, 3-octen-2-one, e, 3e-octen-2-one, 3e-oct-3-en-2-one, hex-1-enyl methyl ketone | |
| (E)-oct-3-en-2-one |
Safety and Handling
TSCA : Yes