missing translation for 'onlineSavingsMsg'
Learn More

3-Octen-2-one 96.0+%, TCI America™

Catalog No. p-7134261
Change view
Click to view available options
Quantity:
25 mL
5 mL
Catalog No. Quantity
O02525ML 5 mL
O025225ML 25 mL
2 options

Catalog No. O02525ML

Supplier: TCI America O02525ML

Only null left

Chemical Identifiers

CAS 1669-44-9
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00015565
InChI Key ZCFOBLITZWHNNC-VOTSOKGWSA-N
Synonym 3-octen-2-one, trans-3-octen-2-one, e-3-octen-2-one, 3e-3-octen-2-one, e-oct-3-en-2-one, unii-m26ah283xv, 3-octen-2-one, e, 3e-octen-2-one, 3e-oct-3-en-2-one, hex-1-enyl methyl ketone
PubChem CID 5363229
IUPAC Name (E)-oct-3-en-2-one
SMILES CCCCC=CC(=O)C

Specifications

Color Yellow
Boiling Point 78°C
Quantity 5 mL
UN Number 1224
Formula Weight 126.20
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Octen-2-one