missing translation for 'onlineSavingsMsg'
Learn More

3-Noradamantanecarboxylic Acid 98.0+%, TCI America™

Catalog No. N06211G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N06211G Supplier TCI America Supplier No. N06211G

Chemical Identifiers

CAS 16200-53-6
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00213463
InChI Key RXUUYFUQAGICCD-NLZKFAMNSA-N
Synonym 3-noradamantanecarboxylic acid, 3-noradamantane carboxylic acid, octahydro-2,5-methanopentalene-3a-carboxylic acid, 2,5-methanopentalene-3a 1h-carboxylic acid, hexahydro, 3-noradamantancarboxylic acid, 3-noradamantanecarboxylicacid, rxuuyfuqagiccd-uhfffaoysa, 3-noradamantanecarboxylate nac, hexahydro-2,5-methano-pentalene-3a-carboxylic acid, tricyclo 3.3.1.0<3,7> nonane-3-carboxylic acid
PubChem CID 11887834
SMILES C1C2CC3CC1CC3(C2)C(=O)O

Specifications

Melting Point 105°C
Color White
Quantity 1 g
Formula Weight 166.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Noradamantanecarboxylic Acid