accessibility menu, dialog, popup

UserName

OO(R)H2C(S)

CAS RN 16200-53-6

IUPAC Name: (1R,5S)-tricyclo[3.3.1.0³,⁷]nonane-3-carboxylate
Synonyms: (1R,5S)-tricyclo[3.3.1.0³,⁷]nonane-3-carboxylate
Molecular Weight (g/mol): 165.21
Molecular Formula: C10H13O2
InChi Key: RXUUYFUQAGICCD-NLZKFAMNSA-M
SMILES: [O-]C(=O)C12C[C@@H]3CC1C[C@H](C2)C3
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results