missing translation for 'onlineSavingsMsg'
Learn More

3-Noradamantanecarboxylic acid, 98%

Catalog No. p-7023287
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Catalog No. Quantity
AAA1668301 0.25 g
AAA1668303 1 g
AAA1668306 5 g
3 options

Catalog No. AAA1668301

Supplier: Thermo Scientific Chemicals A1668301

Only null left

CAS: 16200-53-6 | C10H14O2 | 166.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16200-53-6
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00213463
InChI Key RXUUYFUQAGICCD-NLZKFAMNSA-N
Synonym 3-noradamantanecarboxylic acid, 3-noradamantane carboxylic acid, octahydro-2,5-methanopentalene-3a-carboxylic acid, 2,5-methanopentalene-3a 1h-carboxylic acid, hexahydro, 3-noradamantancarboxylic acid, 3-noradamantanecarboxylicacid, rxuuyfuqagiccd-uhfffaoysa, 3-noradamantanecarboxylate nac, hexahydro-2,5-methano-pentalene-3a-carboxylic acid, tricyclo 3.3.1.0<3,7> nonane-3-carboxylic acid
PubChem CID 11887834
SMILES C1C2CC3CC1CC3(C2)C(=O)O

Specifications

Melting Point 105°C to 107°C
Quantity 0.25 g
Formula Weight 166.22
Percent Purity 98%
Chemical Name or Material 3-Noradamantanecarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only