missing translation for 'onlineSavingsMsg'
Learn More

3-Noradamantanecarboxylic acid, 98%

Catalog No. AAA1668301
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1668301 0.25 g
AAA1668303 1 g
AAA1668306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1668301 Supplier Thermo Scientific Chemicals Supplier No. A1668301
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 16200-53-6 | C10H14O2 | 166.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16200-53-6
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00213463
InChI Key RXUUYFUQAGICCD-NLZKFAMNSA-N
Synonym 3-noradamantanecarboxylic acid, 3-noradamantane carboxylic acid, octahydro-2,5-methanopentalene-3a-carboxylic acid, 2,5-methanopentalene-3a 1h-carboxylic acid, hexahydro, 3-noradamantancarboxylic acid, 3-noradamantanecarboxylicacid, rxuuyfuqagiccd-uhfffaoysa, 3-noradamantanecarboxylate nac, hexahydro-2,5-methano-pentalene-3a-carboxylic acid, tricyclo 3.3.1.0<3,7> nonane-3-carboxylic acid
PubChem CID 11887834
SMILES C1C2CC3CC1CC3(C2)C(=O)O

Specifications

Melting Point 105°C to 107°C
Quantity 0.25 g
Formula Weight 166.22
Percent Purity 98%
Chemical Name or Material 3-Noradamantanecarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only