missing translation for 'onlineSavingsMsg'
Learn More

3-Nonanone 97.0+%, TCI America™

Catalog No. N025325ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N025325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N025325ML Supplier TCI America Supplier No. N025325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 925-78-0
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00009541
InChI Key IYTXKIXETAELAV-UHFFFAOYSA-N
Synonym 3-nonanone, ethyl hexyl ketone, ethyl n-hexyl ketone, n-hexyl ethyl ketone, unii-00c380uhnl, 3-nonanone natural, fema no. 3440, acmc-209rga, octanone, methyl-9ci
PubChem CID 61235
IUPAC Name nonan-3-one
SMILES CCCCCCC(=O)CC

Specifications

Color Yellow
Boiling Point 190°C
Quantity 25 mL
Formula Weight 142.24
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Nonanone